EMAIL THIS PAGE TO A FRIEND
Oncotarget
HDAC inhibitor PAC-320 induces G2/M cell cycle arrest and apoptosis in human prostate cancer.
PMID 29416632
Abstract
HDAC inhibitors (HDACis) have been demonstrated with profound antiproliferative activities in various tumor types. Previously, we screened several polyoxometalate HDACis based on our