EMAIL THIS PAGE TO A FRIEND
Experimental and therapeutic medicine
miR-4262 regulates chondrocyte viability, apoptosis, autophagy by targeting SIRT1 and activating PI3K/AKT/mTOR signaling pathway in rats with osteoarthritis.
PMID 29434702
Abstract
The present study aimed to investigate the effect and underlying mechanism of microRNA (miR)-4262 in the development of osteoarthritis (OA) in rats. Primary chondrocytes were separated from Sprague-Dawley rats and then treated with tumor necrosis factor-α (TNF-α). The level of miR-4262 was detected in TNF-α-treated chondrocytes, and then the miR-4262 or its target gene sirtuin type 1 (SIRT1) level was overexpressed, or knocked down. Furthermore, cell viability, cell apoptosis, cell autophagy and matrix synthesis, as well as the expressions of proteins associated with the phosphoinositide 3-kinase (PI3K)/protein kinase B (AKT)/mammalian target of rapamycin (mTOR) signaling pathway were detected. miR-4262 was significantly overexpressed in TNF-α-treated chondrocytes compared with untreated cells (P<0.05). TNF-α treatment or miR-4262 overexpression significantly decreased cell viability, autophagy-related proteins levels and matrix synthesis-related proteins levels, as well as increased the apoptotic rate in chondrocytes (P<0.05). Overexpression of SIRT1 significantly increased cell viability, autophagy-related proteins levels and matrix synthesis-related proteins levels, as well as decreased the apoptotic rate in TNF-α-treated chondrocytes (P<0.05). In addition, the effects of miR-4262 on cell viability, cell apoptosis, cell autophagy and matrix synthesis were inhibited by SIRT1 (P<0.05). Furthermore, upregulated miR-4262 remarkably increased the expressions of phosphorylated (p)-PI3K, p-AKT and p-mTOR (P<0.05) in TNF-α treated chondrocytes. The present study revealed that the upregulation of miR-4262 may promote the occurrence and development of OA in rats by regulating cell viability, cell apoptosis, cell autophagy, and matrix synthesis. Furthermore, these roles of miR-4262 may be associated with PI3K/AKT/mTOR signaling pathway.