EMAIL THIS PAGE TO A FRIEND
Experimental and therapeutic medicine
L1CAM promotes epithelial to mesenchymal transition and formation of cancer initiating cells in human endometrial cancer.
PMID 29456682
Abstract
Identification of novel factors critical for epithelial to mesenchymal transition (EMT) and cancer initiating cell (CIC) formation may aid in the identification of novel therapeutics for the treatment of endometrial cancer. The present study demonstrated that L1 cell adhesion molecule (CAM) is critical for EMT and formation of CICs in endometrial cancer. Overexpression of L1CAM may promote EMT with increased formation of CICs in HEC-1A endometrial cancer cells. CICs and mesenchymal status resist chemotherapeutic drugs and may regenerate the various cell types in tumors, thereby resulting in relapse of the disease. The present study demonstrated that overexpressing L1CAM promoted paclitaxel resistance and regulated paclitaxel resistance-associated microRNA expression in HEC-1A cells. Furthermore, it was demonstrated that overexpressing L1CAM promoted anoikis resistance in HEC-1A cells. This link between L1CAM and EMT/CICs may provide a novel target for advancing anticancer therapy.