EMAIL THIS PAGE TO A FRIEND
Zhongguo yi xue ke xue yuan xue bao. Acta Academiae Medicinae Sinicae
[Clinical application of the determination of serum digoxin concentration].
PMID 2946462
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L2261
Lanatoside C, ≥95%
C49H76O20