EMAIL THIS PAGE TO A FRIEND
Journal of cosmetic science
Silicone elastomer blends: A novel topical drug delivery platform.
PMID 29465386
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
06753 Isododecane, mixture of isomers, technical, ≥80% (GC)
C12H26