EMAIL THIS PAGE TO A FRIEND
Photodiagnosis and photodynamic therapy
Effective phthalocyanines mediated photodynamic therapy with doxorubicin or methotrexate combination therapy at sub-micromolar concentrations in vitro.
PMID 29476826
Abstract
To improve a cancer patient's quality of life, short treatment duration resulting in rapid tumour removal while sparing normal tissue are highly desirable. Photodynamic therapy (PDT) commonly applied in a single treatment, while often effective can be limited at low photosensitizer or light doses. Combination therapies can overcome the efficacy limitations while not increasing treatment-associated morbidity. Here the efficacy of combination therapy comprised of doxorubicin (DOX) or methotrexate (MTX) with Photosens mediated PDT was investigated in three cell lines in vitro, employing multiple incubation sequences. Photosense is a mixture of aluminium phthalocyanines with different sulfonation. The results demonstrated higher synergistic effects when DOX or MTX-mediated chemotherapy preceded PDT light activation by 24 h. MTX is marginally more cytotoxic than DOX, when combined with Photosens (AlPcS2-4) mediated PDT. While MTX and DOX exposure prior to AlPcS2-4 incubation may enhance mitochondrial localisation photosensitizer, the simultaneous targeting of DNA, proteins, and lipids of the combination therapies leads to the observed high cytotoxicity at sub μM drug doses.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
160415
3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine, ≥99%
C20H14N4