EMAIL THIS PAGE TO A FRIEND
International journal of biological sciences
A Prognostic Model Based on Circulating Tumour Cells is Useful for Identifying the Poorest Survival Outcome in Patients with Metastatic Colorectal Cancer.
PMID 29483832