EMAIL THIS PAGE TO A FRIEND
Contact dermatitis
Contact allergy to EDTA in a topical corticosteroid preparation.
PMID 2948765
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F0891
Flumethasone pivalate
C27H36F2O6