EMAIL THIS PAGE TO A FRIEND
Biochemical pharmacology
Oxidative reactivity of the tryptophan metabolites 3-hydroxyanthranilate, cinnabarinate, quinolinate and picolinate.
PMID 2949752
Abstract
The oxidative reactivities of four tryptophan metabolites in the kynurenine pathway were examined as a potential mechanism for their reported neurotoxicities and carcinogenicities. Neither quinolinic acid, a neurotoxin, nor its monocarboxylic analogue, picolinic acid, auto-oxidized over a wide pH range. However, 3-hydroxyanthranilic acid (3-HAT), a carcinogen, readily auto-oxidized and the reaction rate increased exponentially with increasing pH. 3-HAT auto-oxidation likely involves two steps: auto-oxidation of 3-HAT to the semiquinoneimine (anthranilyl radical) which oxidizes to the quinoneimine, followed by condensation and oxidation reactions to yield a second carcinogen, cinnabarinic acid. 3-HAT auto-oxidation to cinnabarinate required molecular oxygen and generated superoxide radicals and H2O2. Superoxide dismutase (SOD) accelerated 3-HAT auto-oxidation 4-fold, probably by preventing back reactions between superoxide and either the anthranilyl radical or the quinoneimine formed during the initial step of auto-oxidation. Catalase did not accelerate 3-HAT auto-oxidation, but it did prevent destruction of cinnabarinate by H2O2. Interconversion between oxyhemoglobin and methemoglobin occurred during 3-HAT auto-oxidation, although neither form of hemoglobin altered rates of 3-HAT auto-oxidation. Mn2+, Mn3+ and Fe3+-EDTA did not directly catalyze cinnabarinate formation in the absence of O2, but they did accelerate cinnabarinate formation under aerobic conditions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0096
Cinnabarinic Acid, ≥98% (HPLC)
C14H8N2O6