EMAIL THIS PAGE TO A FRIEND
Nihon Jinzo Gakkai shi
[Uremic peak 2a in high performance liquid chromatography--acidic components and their membrane permeability].
PMID 2950263
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
122750
trans-Aconitic acid, 98%
C6H6O6