EMAIL THIS PAGE TO A FRIEND
Journal of mass spectrometry : JMS
Oxidation-assisted structural elucidation of compounds containing a tertiary amine side chain using liquid chromatography mass spectrometry.
PMID 29524298
Abstract
A novel analytical technique for the structural elucidation of compounds bearing a tertiary amine side chain via "in vial" instantaneous oxidation and liquid chromatography mass spectrometry (LC-MS) was developed. A series of lidocaine homologs and benzimidazole derivatives with a major/single amine representative base peak in both their EI-MS and ESI-MS/MS spectra were subjected to oxidation by a 0.1% solution of hydrogen peroxide (including several 16 O/18 O exchange experiments), followed by LC-ESI-MS/MS analysis. The N-oxide counterparts promoted extensive fragmentation with complete coverage of all parts of the molecule, enabling detailed structural elucidation and unambiguous identification of the unoxidized analytes at low nanogram per milliliter levels.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L5647
Lidocaine hydrochloride monohydrate, solid
C14H22N2O · HCl · H2O