EMAIL THIS PAGE TO A FRIEND
Molecular medicine reports
Secretome analysis of rat osteoblasts during icariin treatment induced osteogenesis.
PMID 29532868
Abstract
Osteoporosis is a serious public health problem and icariin (ICA) is the active component of the Epimedium sagittatum, a traditional Chinese medicinal herb. The present study aimed to investigate the effects and underlying mechanisms of ICA as a potential therapy for osteoporosis. Calvaria osteoblasts were isolated from newborn rats and treated with ICA. Cell viability, apoptosis, alkaline phosphatase activity and calcium deposition were analyzed. Bioinformatics analyses were performed to identify differentially expressed proteins (DEPs) in response to ICA treatment. Western blot analysis was performed to validate the expression of DEPs. ICA administration promoted osteoblast viability, alkaline phosphatase activity, calcium deposition and inhibited osteoblast apoptosis. Secretome analysis of ICA‑treated cells was performed using two‑dimensional gel electrophoresis and matrix‑assisted laser desorption/ionization time‑of‑flight mass spectrometry. A total of 56 DEPs were identified, including serpin family F member 1 (PEDF), protein disulfide isomerase family A, member 3 (PDIA3), nuclear protein, co‑activator of histone transcription (NPAT), c‑Myc and heat shock protein 70 (HSP70). These proteins were associated with signaling pathways, including Fas and p53. Bioinformatics and western blot analyses confirmed that the expression levels of the six DEPs were upregulated following ICA treatment. These genes may be directly or indirectly involved in ICA‑mediated osteogenic differentiation and osteogenesis. It was demonstrated that ICA treatment promoted osteogenesis by modulating the expression of PEDF, PDIA3, NPAT and HSP70 through signaling pathways, including Fas and p53.