EMAIL THIS PAGE TO A FRIEND
Zhongguo yi xue ke xue yuan xue bao. Acta Academiae Medicinae Sinicae
[Studies on the synthesis of two new nitrosamines (NMAMBA and NMAMPA) from isoamylamine and sodium nitrite by fungi].
PMID 2953472