EMAIL THIS PAGE TO A FRIEND
The Cochrane database of systematic reviews
Systemic corticosteroids for acute otitis media in children.
PMID 29543327
Abstract
Acute otitis media (AOM) is a common acute infection in children. Pain is its most prominent and distressing symptom. Antibiotics are commonly prescribed for AOM, although they have only a modest effect in reducing pain at two to three days. There is insufficient evidence for benefits of other treatment options, including systemic corticosteroids. However, systemic corticosteroids are potent anti-inflammatory drugs, and so theoretically could be effective, either alone or as an addition to antibiotics. To assess the effects of systemic corticosteroids (oral or parenteral), with or without antibiotics, for AOM in children. We searched the Cochrane Central Register of Controlled Trials (CENTRAL) which contains the Cochrane ARI Group's Specialised Register, MEDLINE (Ovid), Embase (Elsevier), CINAHL (EBSCO), Web of Science (Thomson Reuters), and LILACS (BIREME) for published studies, and ClinicalTrials.gov and the World Health Organization (WHO) International Clinical Trials Registry Platform (ICTRP) for completed and ongoing studies, to 20 February 2018. We checked the reference lists of all primary studies and review articles for additional references and contacted experts in the field to identify additional unpublished materials. We included randomised controlled trials of children with AOM that compared any systemic corticosteroid (oral or parenteral) with placebo, either with antibiotics (corticosteroid plus antibiotic versus placebo plus antibiotic) or without antibiotics (corticosteroid versus placebo). Three review authors (EDS, RR, YP) independently screened the titles and abstracts and retrieved the full texts of potentially relevant studies. We independently extracted study characteristics and outcome data from the included studies, and assessed the risk of bias for each study using the criteria outlined in the Cochrane Handbook for Systematic Reviews of Interventions. We assessed study quality using the GRADE method. We included two studies involving 252 children with AOM aged from three months to six years receiving hospital ambulatory care who were treated with intramuscular ceftriaxone, and who were then randomised to the corticosteroid group (corticosteroid and corticosteroid plus antihistamine) or the placebo group (antihistamine and double placebo). In one study, children also had a needle aspiration of middle ear fluid. Both studies were at unclear risk of bias for allocation concealment, and unclear to high risk of bias for selective reporting.One study (N = 179) included pain as an outcome, but we were unable to derive the proportion of children with persistent pain at Day 5 and Day 14. Reduction of overall or specific symptoms was presented as improvement in clinical symptoms and resolution of inflamed tympanic membranes without the need for additional antibiotic treatment: at Day 5 (94% of children in the treatment group (N = 89) versus 89% in the placebo group (N = 90); risk ratio (RR) 1.06, 95% confidence interval (CI) 0.97 to 1.16) and Day 14 (91% versus 87%; RR 1.05, 95% CI 0.95 to 1.17). Low-quality evidence meant that we are uncertain of the effectiveness of corticosteroids for this outcome.The second study (N = 73) reported a reduction of overall or specific symptoms without additional antibiotic treatment during the first two weeks as a favourable outcome. Children in the treatment group had more favourable outcomes (adjusted odds ratio 65.9, 95% CI 1.28 to 1000; P = 0.037), although the numbers were small. We were unable to pool the results with the other study because it did not report the proportion of children with this outcome by treatment group. Only one study reported adverse effects of corticosteroids (e.g. drowsiness, nappy rash), but did not quantify incidence, so we were unable to draw conclusions about adverse effects. Neither study reported a reduction in overall or specific symptom duration. The evidence for the effect of systemic corticosteroids on AOM is of low to very low quality, meaning the effect of systemic corticosteroids on important clinical outcomes in AOM remains uncertain. Large, high-quality studies are required to resolve the question.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B0385
Beclomethasone, ≥99%
C22H29ClO5