EMAIL THIS PAGE TO A FRIEND
Zhong yao tong bao (Beijing, China : 1981)
[Chemical studies on alkaloids from Veratrum nigrum].
PMID 2954702
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
J4145
Jervine, ≥98% (HPLC), powder
C27H39NO3