EMAIL THIS PAGE TO A FRIEND
Chemosphere
Oxidation of reduced daughter products from 2,4-dinitroanisole (DNAN) by Mn(IV) and Fe(III) oxides.
PMID 29550573
Abstract
Abiotic transformation of anthropogenic compounds by redox-active metal oxides affects contaminant fate in soil. The capacity of birnessite and ferrihydrite to oxidize the insensitive munitions compound, 2,4-dinitroanisol (DNAN), and its amine-containing daughter products, 2-methoxy-5-nitro aniline (MENA) and 2,4-diaminoanisole (DAAN), was studied in stirred reactors at controlled pH (7.0). Aqueous suspensions were reacted at metal oxide solid to solution mass ratios (SSR) of 0.15, 1.5 and 15 g kg-1 and solutions were analyzed after 0-3 h by high performance liquid chromatography coupled with photodiode array or mass spectrometry detection. Results indicate that DNAN was resistant to oxidation by birnessite and ferrihydrite. Ferrihydrite did not oxidize MENA, but MENA was susceptible to rapid oxidation by birnessite, with nitrogen largely mineralized to nitrite. This is the first report on mineralization of nonphenolic aromatics and the release of mineralized N from aromatic amines following reaction with birnessite. DAAN was oxidized by both solids, but ca. ten times higher rate was observed with birnessite as compared to ferrihydrite at an SSR of 1.5 g kg-1. At 15 g kg-1 SSR, DAAN was removed from solution within 5 min of reaction with birnessite. CO2(g) evolution experiments indicate mineralization of 15 and 12% of the carbon associated with MENA and DAAN, respectively, under oxic conditions with birnessite at SSR of 15 g kg-1. The results taken as a whole indicate that initial reductive (bio)transformation products of DNAN are readily oxidized by birnessite. The oxidizability of the reduced DNAN products was increased with progressive (bio)reduction as reflected by impacts on the oxidation rate.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
161195
2-Methoxy-5-nitroaniline, 98%
C7H8N2O3