EMAIL THIS PAGE TO A FRIEND
Zhongguo yao li xue bao = Acta pharmacologica Sinica
[Hypotensive and vasodilator effects of triacetonamine as demonstrated by the intravital microcirculation method].
PMID 2955657