EMAIL THIS PAGE TO A FRIEND
Contact dermatitis
Occupational dermatitis to dihydroxydiphenyl and diphenylthiourea in neoprene gloves.
PMID 2957161
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
168734
4,4′-Dihydroxybiphenyl, 97%
C12H10O2
T28355
N,N′-Diphenylthiourea, 98%
C13H12N2S