EMAIL THIS PAGE TO A FRIEND
Methods in enzymology
Cloning cDNA into lambda gt10 and lambda gt11.
PMID 2958680
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
81268
Poly(ethylene glycol), BioUltra, for molecular biology, 8,000