EMAIL THIS PAGE TO A FRIEND
Advanced materials (Deerfield Beach, Fla.)
A Nonaqueous Potassium-Based Battery-Supercapacitor Hybrid Device.
PMID 29603424
Abstract
A low cost nonaqueous potassium-based battery-supercapacitor hybrid device (BSH) is successfully established for the first time with soft carbon as the anode, commercialized activated carbon as the cathode, and potassium bis(fluoro-slufonyl)imide in dimethyl ether as the electrolyte. This BSH reconciles the advantages of potassium ion batteries and supercapacitors, achieving a high energy density of 120 W h kg-1 , a high power density of 599 W kg-1 , a long cycle life of 1500 cycles, and an ultrafast charge/slow discharge performance (energy density and power density are calculated based on the total mass of active materials in the anode and cathode). This work demonstrates a great potential of applying the nonaqueous BSH for low cost electric energy storage systems.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P11255
PTCDA, 97%
C24H8O6