EMAIL THIS PAGE TO A FRIEND
Bioconjugate chemistry
Protein Surface Structural Recognition in Inactive Areas: A New Immobilization Strategy for Acetylcholinesterase.
PMID 29617563
Abstract
This work reported a new method of design for the immobilization of acetylcholinesterase (AChE) based on its molecular structure to improve its sensitivity and stability. The immobilization binding site on the surface of AChE was determined using MOLCAD's multi-channel functionality. Then, 11 molecules ((+)-catechin, (-)-epicatechin, (-)-gallocatechin, hesperetin, naringenin, quercetin, taxifolin, (-)-epicatechin gallate, flupirtine, atropine, and hyoscyamine) were selected from the ZINC database (about 50 000 molecules) as candidate affinity ligands for AChE. The fluorescence results showed that the binding constant Kb between AChE and the ligands ranged from 0.01344 × 104 to 4.689 × 104 M-1 and there was one independent class of binding site for the ligands on AChE. The AChE-ligand binding free energy ranged from -12.14 to -26.65 kJ mol-1. Naringenin, hesperetin, and quercetin were the three most potent immobilized affinity ligands. In addition, it was confirmed that the binding between the immobilized ligands only occurred at a single site, located in an inactive area on the surface of AChE, and did not affect the enzymatic activity as shown through a competition experiment and enzyme assay. This method based on protein surface structural recognition with high sensitivity and stability can be used as a generic approach for design of the enzyme immobilization and biosensor development.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C1251
(+)-Catechin hydrate, ≥98% (HPLC), powder
C15H14O6 · xH2O
F8927
Flupirtine maleate salt, ≥98% (HPLC)
C15H17FN4O2 · C4H4O4