EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Nicotine induces aberrant hypermethylation of tumor suppressor genes in pancreatic epithelial ductal cells.
PMID 29626481
Abstract
Tobacco smoking is an independent risk factor for the initiation of pancreatic cancer (PC). Hypermethylation of tumor suppressor genes has been demonstrated to be associated with smoking. This study aimed to find the relationship between nicotine exposure and hypermethylation of tumor suppressor genes in normal pancreatic epithelial cells. Human pancreatic epithelial cells ware cultured exposing to nicotine and the methylation status of tumor suppressor genes were detected. Proenkephalin (PENK) was chosen as the target gene and methylation level of PENK promoter region was measured. Expression of DNA methyltransferase (DNMT), nicotine acetylcholine receptor (α7nAChR) and signaling pathway downstream were analyzed. Nicotine induces overexpression of DNMT3A and 3B, and methylated-inactivation of PENK gene in normal pancreatic epithelial cells. An activation of α7nAChR and MAPK signaling pathway has been detected in the nicotine-treated group. Demethylated drug, antagonist of α7nAChR and inhibitor of p38 MAPK is verified to attenuate the overexpression of DNMTs stimulated by nicotine as well as inhibit aberrant hypermethylation-related silence of PENK gene. Nicotine stimulation can induce aberrant hypermethylation of tumor suppressor genes by α7nAChR and MAPK signaling pathway-mediated up-regulation of DNMTs in pancreatic epithelial cells, thus we can provide epigenetic evidence of the mechanisms by which smoking causes pancreatic cancer and find new therapeutic target.