EMAIL THIS PAGE TO A FRIEND
Clinical epigenetics
Competing endogenous RNA expression profiling in pre-eclampsia identifies hsa_circ_0036877 as a potential novel blood biomarker for early pre-eclampsia.
PMID 29643944