EMAIL THIS PAGE TO A FRIEND
Annals of allergy, asthma & immunology : official publication of the American College of Allergy, Asthma, & Immunology
The pediatric asthma yardstick: Practical recommendations for a sustained step-up in asthma therapy for children with inadequately controlled asthma.
PMID 29653238
Abstract
Current asthma guidelines recommend a control-based approach to management involving assessment of impairment and risk followed by implementation of treatment strategies individualized according to the patient's needs and preferences. However, for children with asthma, achieving control can be elusive. Although tools are available to help children (and families) track and manage day-to-day symptoms, when and how to implement a longer-term step-up in care is less clear. Furthermore, treatment is challenged by the 3 age groups of childhood-adolescence (12-18 years old), school age (6-11 years old), and young children (≤5 years old)-and what works for 1 age group might not be the best approach for another. The Pediatric Asthma Yardstick provides an in-depth assessment of when and how to step-up therapy for the child with not well or poorly controlled asthma. Development of this tool follows others in the Yardstick series, presenting patient profiles and step-up strategies based on current guidance documents, but modified according to newer data and the authors' combined clinical experience. The objective is to provide clinicians who treat children with asthma practical and clinically relevant recommendations for each step-up and each intervention, with the intent of helping practitioners better treat their pediatric patients with asthma, particularly those who do not always respond to recommended therapies.