EMAIL THIS PAGE TO A FRIEND
Biochimica et biophysica acta. Gene regulatory mechanisms
Conserved association of Argonaute 1 and 2 proteins with miRNA and siRNA pathways throughout insect evolution, from cockroaches to flies.
PMID 29656113