EMAIL THIS PAGE TO A FRIEND
European heart journal
Safety and efficacy of dual vs. triple antithrombotic therapy in patients with atrial fibrillation following percutaneous coronary intervention: a systematic review and meta-analysis of randomized clinical trials.
PMID 29668889
Abstract
Of patients with atrial fibrillation (AF), approximately 10% undergo percutaneous coronary intervention (PCI). We studied the safety and efficacy of dual vs. triple antithrombotic therapy (DAT vs. TAT) in this population. A systematic review and meta-analysis was conducted using PubMed, Embase, EBSCO, Cochrane database of systematic reviews, Web of Science, and relevant meeting abstracts for Phase 3, randomized trials that compared DAT vs. TAT in patients with AF following PCI. Four trials including 5317 patients were included, of whom 3039 (57%) received DAT. Compared with the TAT arm, Thrombolysis in Myocardial Infarction (TIMI) major or minor bleeding showed a reduction by 47% in the DAT arm [4.3% vs. 9.0%; hazard ratio (HR) 0.53, 95% credible interval (CrI) 0.36-0.85, I2 = 42.9%]. In addition, there was no difference in the trial-defined major adverse cardiac events (MACE) (10.4% vs. 10.0%, HR 0.85, 95% CrI 0.48-1.29, I2 = 58.4%), or in individual outcomes of all-cause mortality, cardiac death, myocardial infarction, stent thrombosis, or stroke between the two arms. Compared with TAT, DAT shows a reduction in TIMI major or minor bleeding by 47% with comparable outcomes of MACE. Our findings support the concept that DAT may be a better option than TAT in many patients with AF following PCI.