EMAIL THIS PAGE TO A FRIEND
Arthritis & rheumatology (Hoboken, N.J.)
Toll-Like Receptor 9 Deficiency Breaks Tolerance to RNA-Associated Antigens and Up-Regulates Toll-Like Receptor 7 Protein in Sle1 Mice.
PMID 29687651
Abstract
Toll-like receptors (TLRs) 7 and 9 are important innate signaling molecules with opposing roles in the development and progression of systemic lupus erythematosus (SLE). While multiple studies support the notion of a dependency on TLR-7 for disease development, genetic ablation of TLR-9 results in severe disease with glomerulonephritis (GN) by a largely unknown mechanism. This study was undertaken to examine the suppressive role of TLR-9 in the development of severe lupus in a mouse model. We crossed Sle1 lupus-prone mice with TLR-9-deficient mice to generate Sle1TLR-9-/- mice. Mice ages 4.5-6.5 months were evaluated for severe autoimmunity by assessing splenomegaly, GN, immune cell populations, autoantibody and total Ig profiles, kidney dendritic cell (DC) function, and TLR-7 protein expression. Mice ages 8-10 weeks were used for functional B cell studies, Ig profiling, and determination of TLR-7 expression. Sle1TLR-9-/- mice developed severe disease similar to TLR-9-deficient MRL and Nba2 models. Sle1TLR-9-/- mouse B cells produced more class-switched antibodies, and the autoantibody repertoire was skewed toward RNA-containing antigens. GN in these mice was associated with DC infiltration, and purified Sle1TLR-9-/- mouse renal DCs were more efficient at TLR-7-dependent antigen presentation and expressed higher levels of TLR-7 protein. Importantly, this increase in TLR-7 expression occurred prior to disease development, indicating a role in the initiation stages of tissue destruction. The increase in TLR-7-reactive immune complexes, and the concomitant enhanced expression of their receptor, promotes inflammation and disease in Sle1TLR9-/- mice.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
RAB0806 Mouse IgM ELISA Kit