EMAIL THIS PAGE TO A FRIEND
The Journal of Biological Chemistry
Ankyrin repeat and Single KH domain 1 (ANKHD1) drives renal cancer cell proliferation via binding to and altering a subset of miRNAs.
PMID 29695508