PloS one
Effects of isomaltodextrin in postprandial lipid kinetics: Rat study and human randomized crossover study.
PMID 29715296
Abstract
Isomaltodextrin (IMD) is a novel dietary fiber-like polysaccharide: a type of α-glucan produced from starch using enzymes derived from microorganisms. The results of cohort studies show that dietary fiber can prevent cardiovascular disorders caused by lifestyle-related diseases such as metabolic syndrome. Inhibition of excess fat absorption by dietary fiber is known to be one of the mechanisms, and it is also known that the actions of dietary fiber vary depending on factors such as its structure or origin. Thus, we investigated the inhibitory actions of IMD on fat absorption, and analyzed its mechanism of action. In rats, the absorption of fat given by gavage was significantly lower at 1, 2, and 6 hours after IMD administration than after vehicle administration. In humans, IMD was associated with a lesser increase in blood triglycerides in subjects whose blood triglycerides were otherwise apt to rise. We also found by in vitro emulsion studies that IMD, which had no effect on digestive enzyme activity or emulsion formation, stabilized the micro size micelle by inducing enlarged micelle particle size and increased zeta potential. In conclusion, the mechanism of inhibition of fat absorption by IMD may be a delay in micelle particles accessing the intestinal epithelium through changes in the surface structure and the physical properties of the micelle particles.