EMAIL THIS PAGE TO A FRIEND
Journal of the American Heart Association
Gene Transfer of Engineered Calmodulin Alleviates Ventricular Arrhythmias in a Calsequestrin-Associated Mouse Model of Catecholaminergic Polymorphic Ventricular Tachycardia.
PMID 29720499
Abstract
Catecholaminergic polymorphic ventricular tachycardia (CPVT) is a familial arrhythmogenic syndrome characterized by sudden death. There are several genetic forms of CPVT associated with mutations in genes encoding the cardiac ryanodine receptor (RyR2) and its auxiliary proteins including calsequestrin (CASQ2) and calmodulin (CaM). It has been suggested that impairment of the ability of RyR2 to stay closed (ie, refractory) during diastole may be a common mechanism for these diseases. Here, we explore the possibility of engineering CaM variants that normalize abbreviated RyR2 refractoriness for subsequent viral-mediated delivery to alleviate arrhythmias in non-CaM-related CPVT. To that end, we have designed a CaM protein (GSH-M37Q; dubbed as therapeutic CaM or T-CaM) that exhibited a slowed N-terminal Ca dissociation rate and prolonged RyR2 refractoriness in permeabilized myocytes derived from CPVT mice carrying the CASQ2 mutation R33Q. This T-CaM was introduced to the heart of R33Q mice through recombinant adeno-associated viral vector serotype 9. Eight weeks postinfection, we performed confocal microscopy to assess Ca handling and recorded surface ECGs to assess susceptibility to arrhythmias in vivo. During catecholamine stimulation with isoproterenol, T-CaM reduced isoproterenol-promoted diastolic Ca waves in isolated CPVT cardiomyocytes. Importantly, T-CaM exposure abolished ventricular tachycardia in CPVT mice challenged with catecholamines. Our results suggest that gene transfer of T-CaM by adeno-associated viral vector serotype 9 improves myocyte Ca handling and alleviates arrhythmias in a calsequestrin-associated CPVT model, thus supporting the potential of a CaM-based antiarrhythmic approach as a therapeutic avenue for genetically distinct forms of CPVT.