EMAIL THIS PAGE TO A FRIEND
Calcified tissue international
Glycyrrhizin Suppresses RANKL-Induced Osteoclastogenesis and Oxidative Stress Through Inhibiting NF-κB and MAPK and Activating AMPK/Nrf2.
PMID 29721581
Abstract
The treatment for osteoporosis involves inhibiting bone resorption and osteoclastogenesis. Glycyrrhizin (GLY) is a triterpenoid saponin glycoside known to be as the most medically efficacious component of the licorice plant. It has strong anti-inflammatory, antioxidant, and antitumor properties. We investigated the effect of GLY on osteoclastogenesis, bone resorption, and intracellular oxidative stress and its molecular mechanisms. In vitro osteoclastogenesis assays were performed using bone marrow monocytes with and without glycyrrhizin. We also evaluated the effects of glycyrrhizin on the secretion of TNF-α, IL-1β, and IL-6 in LPS-stimulated RAW 264.7 cells using ELISA. The effects of glycyrrhizin on the expression of osteoclast-related genes, such as Nfatc1, c-fos, Trap, and cathepsin K (CK), were investigated by RT-PCR. Intracellular reactive oxygen species (ROS) were detected in receptor activator of nuclear factor kappa-Β ligand (RANKL)-stimulated osteoclasts in the presence and absence of glycyrrhizin. During the inhibition of osteoclastogenesis by glycyrrhizin, phosphorylation of AMPK, Nrf2, NF-κB, and MAPK was analyzed using western blotting. Our results showed that glycyrrhizin significantly inhibited RANKL-induced osteoclastogenesis, downregulated the expression of NFATc1, c-fos, TRAP, CK, DC-STAMP, and OSCAR, and inhibited p65, p38, and JNK. Glycyrrhizin was found to significantly decrease the secretion of inflammatory cytokines (TNF-α, IL-1β, and IL-6). Additionally, glycyrrhizin reduced the formation of ROS in osteoclasts by inducing AMPK phosphorylation and nuclear transfer of NRF2, resulting in an upregulation of antioxidant enzymes, such as HO-1, NQO-1, and GCLC. In summary, we found that glycyrrhizin inhibited RANKL-induced osteoclastogenesis. It was also indicated that glycyrrhizin could reduce oxidative stress by inhibiting the MAPK and NF-κB pathways and activating the AMPK/NRF2 signaling. Therefore, glycyrrhizin may be used as an effective therapeutic agent against osteoporosis and bone resorption.