EMAIL THIS PAGE TO A FRIEND
Contact dermatitis
Dermatitis from crystal violet.
PMID 2972522
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
332488
Crystal Violet lactone, Dye content 97 %
C26H29N3O2