EMAIL THIS PAGE TO A FRIEND
Scientific reports
2H-NbS2 film as a novel counter electrode for meso-structured perovskite solar cells.
PMID 29728660
Abstract
We report the use of 2H-NbS2 film as a novel counter electrode in perovskite solar cells fabricated with a cold isostatic pressing method. The 2H-NbS2 film, which was prepared through an exfoliation method followed by restacking from LixNbS2 powder, shows high electrical conductivity of 8.7 × 103 S cm-1 and work function of 5.20 eV. The two-dimensional transition metal dichalcogenide was used for the first time as a counter electrode in meso-structured perovskite solar cells. Through this process, we demonstrated a new alternative to noble metals. The perovskite solar cell base on the 2H-NbS2 counter electrode showed an open-circuit voltage of 1.046 V, comparable to that of gold, and a power conversion efficiency of 8.3%.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
142379
4-tert-Butylpyridine, 98%
C9H13N