PloS one
Phytochemical study of the headspace volatile organic compounds of fresh algae and seagrass from the Adriatic Sea (single point collection).
PMID 29738535
Abstract
Performed phytochemical study contributes to the knowledge of volatile organic compounds (VOCs) of Halopteris filicina (Grateloup) Kützing, Dictyota dichotoma (Hudson) J. V. Lamouroux, Posidonia oceanica (L.) Delile and Flabellia petiolata (Turra) Nizamuddin from the Adriatic Sea (single point collection). VOCs were investigated by headspace solid-phase microextraction (HS-SPME) and analysed by gas chromatography and mass spectrometry (GC-MS/FID). H. filicina headspace contained dimethyl sulfide (DMS; 12.8%), C8-compounds (e.g. fucoserratene (I; 9.5%)), benzaldehyde (II; 8.7%), alkane C17, dictyopterene D and C (III, IV), tribromomethane (V), 1-iodopentane, others. F. petiolata headspace was characterized by DMS (22.2%), 6-methylhept-5-en-2-one (9.5%), C17 (9.1%), II (6.5%), compounds I-V. DMS (59.3%), C15 (14.5%), C17 (7.2%) and C19 (6.3%) dominated in P. oceanica headspace. Sesquiterpenes were found in D. dichotoma, predominantly germacrene D (28.3%) followed by other cadinenyl (abundant), muurolenyl and amorphenyl structures. Determined VOCs may be significant for chemosystematics and chemical communications in marine ecosystem.