EMAIL THIS PAGE TO A FRIEND
Journal of applied microbiology
Antibacterial potential of phytochemicals alone or in combination with antimicrobials against fish pathogenic bacteria.
PMID 29741243
Abstract
This study investigated the antibacterial activity of five phytochemicals (carvacrol, citral, eugenol, linalool and thymol) alone or in combination with florfenicol or oxytetracycline against bacteria isolated from silver catfish (Rhamdia quelen). We also analysed the potential of these compounds to inhibit biofilm formation and haemolysis caused by the bacteria. Bacteria were tested with antimicrobials to calculate the multiple antibiotic resistances. The checkerboard assay was used to evaluate a putative synergy between five phytochemicals and antimicrobials against the strains isolated. The biofilm formation inhibition assay was performed with phytochemicals and antimicrobials, and the haemolysis inhibition assay was performed with the phytochemicals. Carvacrol, eugenol and thymol were the most effective phytochemicals. Three combinations (linalool with florfenicol or oxytetracycline against Aeromonas hydrophila and citral with oxytetracycline against Citrobacter freundii) demonstrated synergy in the checkerboard assay. All phytochemicals inhibited biofilm formation and haemolysis activity. The tested phytochemicals showed satisfactory activity against fish pathogenic bacteria. The phytochemicals did not present antagonistic interactions with the antimicrobials, allowing their combined use, which may contribute to a decrease in the use of conventional drugs and their residues in aquatic environment.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F1427
Florfenicol, analytical standard, for drug analysis
C12H14Cl2FNO4S