EMAIL THIS PAGE TO A FRIEND
Advances in alcohol & substance abuse
Discovery of novel brain lipoxygenase products formed from docosahexaenoic acid (22:6w3).
PMID 2975919
Abstract
It is known that alcohol exposure leads to a decline in brain docosahexaenoate (22:6w3). We hypothesized that alcohol could stimulate the metabolism of this polyunsaturated fatty acid to bioactive products. Several oxidized products of 22:6w3 were indeed observed when rat brain homogenate was incubated with 14C22:6w3 in vitro. A similar group of metabolites was formed in vivo from 14C-22:6w3 injected into the lateral ventricle. These metabolites were characterized by thermospray- and GC/MS as well as by the synthesis of standards using purified enzymes. Platelet lipoxygenase also proved useful in identifying one of the brain metabolites and served as a source of enzyme for preparative studies. Their physiological effects on smooth muscle tone and platelet aggregation will be presented.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D2534
cis-4,7,10,13,16,19-Docosahexaenoic acid, ≥98%
C22H32O2