EMAIL THIS PAGE TO A FRIEND
Phytochemistry
High correlation of chemical composition with genotype in cryptic species of the liverwort Aneura pinguis.
PMID 29763782
Abstract
Chemical constituents of cryptic species detected within the liverwort Aneura pinguis were identified using headspace solid-phase microextraction gas chromatography-mass spectrometry (HS-SPME-GC-MS). Fibre coating with divinylbenzene/carboxen/polydimethylsiloxane (DVB/CAR/PDMS) was used. A total of 48 samples of A. pinguis were analysed. The studied plants were identified genetically based on barcode DNA sequences and represented three cryptic species (A, B and F) of A. pinguis. Cryptic species A and B are genetically diverse; both represent three evolutionary lineages: A1, A2, A3 and B1, B2, B3, respectively. The cryptic species F that was recently detected is not diverse. The most characteristic compounds in analysed samples were sesquiterpene hydrocarbons (up to 17.7% for A1; 15.7% for A2; 20.6% for A3; 7.7% for B1; 2.0% for B2; 3.7% for B3; 10.2% for F), oxygenated sesquiterpenoids (up to 68.0% for A1; 54.7% for A2; 52.6% for A3; 63.5% for B1; 88.7% for B2; 82.7% for B3; 78.8% for F), and linear aliphatic hydrocarbons (up to 14.8% for A1; 1.1% for A2; 12.1% for A3; 6.9 for B1; 5.2% for B2; 1.1% for B3; 7.0% for F). The dominant compound in the studied samples was pinguisone. The second dominant compound present in the tested plant material was deoxopinguisone, except for lineage B2, where only a small relative concentration of this compound was found. A high content of deoxopinguisone in cryptic species A (lineages A1, A2 and A3) was accompanied by the presence of isopinguisone and methyl norpinguisonate, whereas these two compounds were not detected in cryptic species B (lineages B1 and B3) and F. The chemical compounds detected in the studied samples of A. pinguis were subjected to multivariate statistical analysis. The results showed that the chemical composition depends mainly on the genotype of the plant and slightly on the habitat. However, there was no clear correlation between the volatile compounds and the date of collection of the studied plants.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
25461 Carboxen®-1006 PLOT Capillary GC Column, L × I.D. 30 m × 0.53 mm, average thickness 30 μm
24241-U Carboxen®-1006 PLOT Capillary GC Column, L × I.D. 30 m × 0.32 mm, average thickness 15 μm
25467 Carboxen®-1010 PLOT Capillary GC Column, L × I.D. 30 m × 0.53 mm, average thickness 30 μm
24246 Carboxen®-1010 PLOT Capillary GC Column, L × I.D. 30 m × 0.32 mm, average thickness 15 μm
54041-U Carboxen® 1000 Reversible SPE Tube, 200 mg/mL, pk 30
29753-U Carboxen® 569, O.D. × L 1/4  in. × 3 1/2 , glass TD tube, fritted, Sealed with Brass Endcaps, preconditioned, pkg of 10 ea
29534-U Carboxen® 569, glass TD tube, fritted, O.D. × L 1/4  in. × 3 1/2  in., preconditioned, pkg of 10 ea
54293-U Carboxen® 572 for Tobacco Smoke, Adsorbent Bed Wt: (300 mg), cartridge volume 3 mL, pkg of 10 ea
54294-U Carboxen® 572 for Tobacco Smoke, Adsorbent Bed Wt: 300 mg, cartridge size 3 mL, pkg of 50 ea
10478-U Carboxen® Adsorbent, matrix Carboxen® 1000, 60-80 mesh, bottle of 10 g
11048-U Carboxen® Adsorbent, matrix Carboxen® 569, 20-45 mesh, bottle of 500 g
10477-U Carboxen® Adsorbent, matrix Carboxen® 1000, 40-60 mesh, bottle of 50 g
11324-U Carboxen® Adsorbent, matrix Carboxen® 564, 20-45 mesh, pack of 144 × 290 mg
11072-U Carboxen® Adsorbent, matrix Carboxen® 572, 20-45 mesh, bottle of 10 g
23446-U ORBO-93 Sorbent Tube, matrix Carboxen® 1000, ORBO tube I.D. × L 6 mm × 95 mm, Bed B 90 mg, Bed A 180 mg, 60/80 mesh, pkg of 25 ea
20355 ORBO 78 HBr on Carboxen® 564 specially cleaned (20/45), 400/200 mg, W,W,W separators, O.D. × L 6 mm × 110 mm, pkg of 25 ea
20358 ORBO 90 Carboxen® 564 (20/45), 160/80 mg, W,F,F separators, O.D. × L 6 mm × 75 mm, pkg of 25 ea, for analyte group MEK (methylethyl ketone)
28718-U Tenax® TA / Carboxen® 1018, glass TD tube, preconditioned, O.D. × L 1/4  in. × 3 1/2  in., Sealed with (Swagelok® End-Fittings), pkg of 10 ea