EMAIL THIS PAGE TO A FRIEND
Gigiena i sanitariia
[Hygienic standardization of isobutyronitrile in reservoir water].
PMID 2976707
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
538248
Isobutyronitrile, 99.6%
C4H7N
I15602
Isobutyronitrile, 99%
C4H7N