EMAIL THIS PAGE TO A FRIEND
Gigiena truda i professional'nye zabolevaniia
[Effectiveness of introducing individual protection measures for workers in the phenazone industry].
PMID 2977372
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45385
Chloridazon, PESTANAL®, analytical standard
C10H8ClN3O