EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Extracellular matrix protein DMP1 suppresses osteogenic differentiation of Mesenchymal Stem Cells.
PMID 29775615
Abstract
Mesenchymal Stem Cells (MSCs) are self-renewing and multipotent stem cells which was investigated for diverse clinical applications. However, complex mechanism of MSCs fate determination is still not fully disclosed. Extracellular matrix (ECM) proteins contribute to maintain MSCs stemness by providing extracellular microenvironment. Increasing evidences show that ECM proteins could also regulate the fate of MSCs directly. Dentin matrix protein 1 (DMP1) is an ECM protein enrich in bone tissue and terminal cells, which well-known in promoting osteoblasts and osteocytes maturation, and facilitate mineralization. Recently, our experiment indicated that DMP1 was also expressed in MSCs of long bone. In present study, it is found that DMP1 expressed in Prx1 positive MSCs. And, DMP1 is down-regulated in early osteoblasts and up-regulated again in mature osteoblasts. DMP1 conditional knockout mice model under Prx1cre was generated to explore whether DMP1 regulates MSCs osteogenic differentiation. Specific ablation of DMP1 in Prx1 positive MSCs increased bone mass in vivo and promoted osteoblasts activity in vitro. This study provides a new understanding of DMP1's function in regulation of osteogenesis: not only an enhancer of bone formation, but also a negative regulator of MSCs differentiation in bone.