EMAIL THIS PAGE TO A FRIEND
Acta histochemica
Anatomical, histological and immunohistochemical study of testicular development in Columba livia (Aves: Columbiformes).
PMID 29776745
Abstract
In this work, testicular ontogeny is analyzed at the anatomical, histological and immunohistochemical levels; the latter through the detection of GnRHR and PCNA in the testicles of embryos, neonates and juveniles of Columba livia. We analyzed 150 embryos, 25 neonates and 5 juveniles by means of observations under a stereoscopic magnifying glass and scanning electron microscope (SEM). The histological analysis was performed using hematoxylin-eosin staining techniques and the PAS reaction. For the immunohistochemical analysis, the expression of GnRHR and PCNA in embryos corresponding to stages 41, 43 and in neonates of 2, 5, 7 and 75 days post-hatch was revealed in testicular histological preparations. That gonadal outline is evident in stage 18. In stage 29, the testes are constituted of a medulla in which the PGCs are surrounded by the Sertoli cells, constituting the seminiferous tubules. From stage 37 a greater organization of the tubules is visualized and at the time of hatching the testicle is constituted of the closed seminiferous tubules, formed of the PGCs and Sertoli cells. The Leydig cells are evident outside the tubules. In the juvenile stages, the differentiation of germline cells and the organization of small vessels that irrigate the developing testicle begin to be visible. In the analyzed stages, the immunodetection of the GnRHR receptor and PCNA revealed specific marking in the plasma membrane and in the perinuclear zone for GnRHR and in the nucleus of the germline cells in juvenile testicles for PCNA. These results can be used as a basis for further study of endocrine regulation events during testicular ontogeny in avian species.