EMAIL THIS PAGE TO A FRIEND
Methods (San Diego, Calif.)
Screening and selection of artificial riboswitches.
PMID 29778645
Abstract
Synthetic riboswitches are engineered to regulate gene expression in response to a variety of non-endogenous small molecules, and a challenge to select this engineered response requires robust screening tools. A new synthetic riboswitch can be created by linking an in vitro-selected aptamer library with a randomized expression platform followed by in vivo selection and screening. In order to determine response to analyte, we developed a dual-color reporter comprising elements of the E. coli fimbriae phase variation system: recombinase FimE controlled by a synthetic riboswitch and an invertible DNA segment (fimS) containing a constitutively active promoter placed between two fluorescent protein genes. Without an analyte, the fluorescent reporter constitutively expressed green fluorescent protein (GFPa1). Addition of the analyte initiated translation of fimE causing unidirectional inversion of the fimS segment and constitutive expression of red fluorescent protein (mKate2). The dual color reporter system can be used to select and to optimize artificial riboswitches in E. coli cells. In this work, the enriched library of aptamers incorporated into the riboswitch architecture reduces the sequence search space by offering a higher percentage of potential ligand binders. The study was designed to produce structure switching aptamers, a necessary feature for riboswitch function and efficiently quantify this function using the dual color reporter system.
Related Materials