EMAIL THIS PAGE TO A FRIEND
Molecular nutrition & food research
Theobromine Does Not Affect Fasting and Postprandial HDL Cholesterol Efflux Capacity, While It Decreases Fasting miR-92a Levels in Humans.
PMID 29797695
Abstract
Chocolate consumption lowers cardiovascular disease risk, which might be attributed to the methylxanthine theobromine. These effects may be mediated through effects on HDL-mediated cholesterol efflux, which may be affected by microRNA (miRNA) levels in the HDL particles. Therefore, the aim of this study is to investigate effects of theobromine consumption on fasting and postprandial cholesterol efflux and miRNAs levels. Thirty overweight and 14 obese healthy men and women participated in this randomized, double-blind crossover study. Participants consumed 500 mg d-1 of theobromine or placebo for 4 weeks. ABCA1-mediated cholesterol efflux was measured using J774 macrophages. MiRNAs levels (miR-92a, miR-223, miR-135a*) were quantified in apolipoprotein B-depleted serum. Theobromine consumption did not affect fasting and postprandial cholesterol efflux. Fasting miR-223 and miR-135a levels were unchanged, while miR-92a levels were decreased (-0.21; p < 0.05). The high-fat meal increased postprandial cholesterol efflux capacity (+4.3 percentage points; p ≤ 0.001), miR-92a (+1.21; p < 0.001), and miR-223 (+1.79; p < 0.001) levels, while a trend was found for miR-135a (+1.08; p = 0.06). Theobromine did not improve fasting and postprandial ABCA1-mediated cholesterol efflux capacity, but decreased fasting miR-92a levels. High-fat meal intake increased postprandial cholesterol efflux and the three selected miRNAs levels.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S9318
Sandoz 58-035, >98% (HPLC), powder
C30H47NOSi