EMAIL THIS PAGE TO A FRIEND
The Journal of the Egyptian Public Health Association
Hepatotoxicity of some antimoulting compounds in white male mice.
PMID 2979938
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
36530
Chlorfluazuron, PESTANAL®, analytical standard
C20H9Cl3F5N3O3