EMAIL THIS PAGE TO A FRIEND
Histology and histopathology
Effects of mellitic acid (MA) and sodium fluoride (NaF) on the histological appearance of murine fetal tibiae cultured in vitro.
PMID 2980123
Abstract
The aim of this study was to develop a standardized image analysis method for localization and quantitative measurement of calcified structures of murine fetal tibiae cultured in vitro as a completion and verification of previous biochemical studies. The calcified structures of bone stained by von Kossa silver technique and the epiphyseal cartilages showing intensive metachromasia with toluidine-blue staining were converted with grey-value window programs and afterwards the areas of the selected structures were measured. The histomorphological investigations showed that the murine tibiae, incubated for a period of 6 days in a medium with addition of 5 mmol mellitic acid, showed both a significant reduction of calcium deposits and an increase of epiphyseal intercellular cartilage matrix. The tibiae incubated in a medium with addition of 0.5 mmol sodium fluoride significantly showed an increase of calcium deposits in the thickened lamellae of the compacta. These histomorphological results confirm previous biochemical studies.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M2705
Mellitic acid, 99%
C12H6O12