EMAIL THIS PAGE TO A FRIEND
Experientia
The use of indentometry to study the effect of agents known to increase skin c-AMP content.
PMID 2982648
Abstract
Local, externally applied pharmacological agents which are assumed to raise the c-AMP level, decrease the low pressure indentation value of the forehead skin of certain human volunteers.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I2760
(−)-Isoproterenol (+)-bitartrate salt, powder
C11H17NO3 · C4H6O6