EMAIL THIS PAGE TO A FRIEND
Experimental and therapeutic medicine
Assessment of efficacy of prenatal genetic diagnosis for fragile X syndrome using nested PCR.
PMID 29844802
Abstract
Fragile X syndrome (FXS) is the most common inherited cause of intellectual disability and the leading monogenic cause of autism spectrum disorder. It has previously been demonstrated that prenatal genetic diagnosis is efficient for the diagnosis of FXS. The present study investigated the diagnostic effects of nested polymerase chain reaction (PCR) for fragile X mental retardation 1 (FMR1) and expanded CGG repeats. It was demonstrated that the nested PCR assay rapidly measured the multi-copies of the FMR1 gene in individual samples. The nested PCR assay detected normal CGG repeat lengths and expanded CGG repeat lengths with a low occurrence of false positives. In addition, the nested PCR assay resulted in increased sensitivity and specificity for patients with FXS. Furthermore, the nested PCR assay identified the mutation and generated conclusive cases for FXS, indicating that this assay is beneficial for the diagnosis of FXS patients. In conclusion, these outcomes indicate that nested PCR assay is a reliable and easier method for diagnosis of FXS, which may be used for the diagnosis of FXS patients.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D8783
7-Deaza-2′-deoxyguanosine 5′-triphosphate lithium salt, 10 mM in H2O
C11H17N4O13P3