EMAIL THIS PAGE TO A FRIEND
Aquatic toxicology (Amsterdam, Netherlands)
Dose- and age-specific antioxidant responses of the mysid crustacean Neomysis awatschensis to metal exposure.
PMID 29859404
Abstract
Waterborne metals can adversely affect an organism's innate defenses through oxidative stress. In the present study, the marine mysid Neomysis awatchensis was exposed to sublethal concentrations (1/10 and 1/5 of the median LC50s) of As, Cd, Cu, Pb, and Zn for 48 or 96 h at the juvenile and adult developmental stages, and the dose- and age-specific antioxidant defense system responses were characterized. Metal accumulation and modulation of four key antioxidant biomarkers, malondialdehyde (MDA), glutathione (GSH), superoxide dismutase, and catalase, were measured in juvenile and adult mysids. Based on LC50, level of metal toxicity was dependent on metal concentration, developmental stage, and the exposure duration. Intracellular MDA content was increased in the As-, Cu-, Pb-, and Zn-exposed juvenile mysids after exposure for 48 and/or 96 h, while increases in MDA content were observed in adult mysids following Cu and Zn exposure. Interestingly, GSH content was differentially modulated, where intracellular GSH levels decreased in juvenile mysids following Cd, Cu, Pb, and Zn exposure, but significantly increased in metal-exposed adult mysids, except for Pb exposure. The catalase and superoxide dismutase activities displayed similar stage-specific increases or decreases as also observed for the different GSH levels, suggesting that the susceptibility to and defense against metal-induced oxidative stress differed based on stage. Modulations in MDA and GSH content and enzymatic activity of the antioxidant defense system indicate that mysid antioxidant defense system factors are intimately connected during control of oxidative imbalances with different capacities at different developmental stages.