EMAIL THIS PAGE TO A FRIEND
Development (Cambridge, England)
PDGF-A suppresses contact inhibition during directional collective cell migration.
PMID 29884673
Abstract
The leading-edge mesendoderm (LEM) of the Xenopus gastrula moves as an aggregate by collective migration. However, LEM cells on fibronectin in vitro show contact inhibition of locomotion by quickly retracting lamellipodia upon mutual contact. We found that a fibronectin-integrin-syndecan module acts between p21-activated kinase 1 upstream and ephrin B1 downstream to promote the contact-induced collapse of lamellipodia. To function in this module, fibronectin has to be present as puncta on the surface of LEM cells. To overcome contact inhibition in LEM cell aggregates, PDGF-A deposited in the endogenous substratum of LEM migration blocks the fibronectin-integrin-syndecan module at the integrin level. This stabilizes lamellipodia preferentially in the direction of normal LEM movement and supports cell orientation and the directional migration of the coherent LEM cell mass.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M2786
3-Maleimidobenzoic acid N-hydroxysuccinimide ester, crystalline
C15H10N2O6