EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
LRP4 promotes proliferation, migration, and invasion in papillary thyroid cancer.
PMID 29885843
Abstract
Dysregulation of cell proliferation and death is considered the foundation of the malignant biological characteristics of cancer. In this study, we conducted a comprehensive analysis of a massively parallel whole transcriptome resequencing of paired papillary thyroid cancer and normal thyroid tissues from 19 patients. In addition, we found that LRP4, a member of the low-density lipoprotein receptor-related protein family, is significantly overexpressed in thyroid carcinoma. We demonstrated through quantitative real-time polymerase chain reaction (qRT-PCR) that LRP4 is upregulated in papillary thyroid cancer (PTC) tissues. This observation was also consistent with data analyzed from The Cancer Genome Atlas (TCGA) cohort. Thus, the biological role of LRP4 in the thyroid cancer in the present study was investigated using the PTC cell lines TPC1, BCPAP and KTC-1. In these cell lines, the mRNA level of LRP4 was higher than normal thyroid cancer cell named HTORI3. In vitro experiments demonstrated that LRP4 downregulation significantly inhibited the colony formation, proliferation, migration, and invasion of the three PTC cell lines. Knockdown of LRP4 by small interfering RNA (siRNA) in those cell lines decreased the protein expression of N-cadherin, Enhancer of zeste homolog 2 (EZH2), and Zinc finger E-box-binding home-box 1 (ZEB1). Furthermore, LRP4 knockdown significantly reduced the levels of phosphorylated PI3K in the PTC cell lines. In conclusion, the present study indicated that LRP4 is a gene associated with PTC and might become a potential therapeutic target.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EHU017401 MISSION® esiRNA, esiRNA targeting human LRP4 (esiRNA1)